Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276503.1 | NP_001263432.1 | 3 | 359 | ||
NM_001276504.1 | NP_001263433.1 | 3 | 387 | |||
NM_001276506.1 | NP_001263435.1 | 5 | 634 | |||
NM_003002.3 | NP_002993.1 | 4 | 504 | |||
NR_077060.1 | 5 | 558 | ||||
GenBank mRNA | AB006202.1 | - | 4 | 431 | ||
AK075360.1 | - | 4 | 430 | |||
BC005263.1 | - | 4 | 448 | 3930691 | ||
BC009574.1 | - | 4 | 429 | 4063808 | ||
BC012603.1 | - | 4 | 433 | 4108104 | ||
BC015188.1 | - | 4 | 455 | 4279214 | ||
BC015992.1 | - | 4 | 451 | 4292983 | ||
BC022350.1 | - | 4 | 443 | 4275516 | ||
BC070307.1 | - | 4 | 429 | 4277540 | ||
BC071755.1 | - | 4 | 447 | 4774525 | ||
BC071756.1 | - | 4 | 429 | 6450992 | ||
BE567994.1 | - | 5 | 451 | |||
BF211358.1 | - | 3 | 312 | |||
BQ932668.1 | - | 4 | 512 | |||
BT007238.1 | - | 4 | 420 | |||
CD105572.1 | - | 3 | 299 | |||
CR456932.1 | - | 4 | 420 | |||
KU178325.1 | - | 4 | 420 | |||
KU178326.1 | - | 3 | 275 |