Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC085271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC085271.1 | - | 1 | 569 | 30146389 |
Gene Symbol: Set | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204875.1 | NP_001191804.1 | 5 | 739 | ||
NM_023871.4 | NP_076360.1 | 5 | 604 | |||
GenBank mRNA | AB015613.1 | - | 5 | 403 | ||
AB044937.1 | - | 5 | 488 | |||
AK019960.1 | - | 5 | 488 | |||
AK172327.1 | - | 5 | 748 | |||
BC018255.1 | - | 5 | 557 | 4014298 | ||
BC103791.1 | - | 5 | 432 | 6511600 |