Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009214.3 | NP_033240.3 | 7 | 776 | ||
GenBank mRNA | AF031486.1 | - | 7 | 759 | ||
AK017775.1 | - | 7 | 747 | |||
AK049780.1 | - | 6 | 1534 | |||
AK049787.1 | - | 7 | 765 | |||
AK136798.1 | - | 7 | 777 | |||
AK149929.1 | - | 7 | 774 | |||
AK160659.1 | - | 7 | 777 | |||
AK166665.1 | - | 7 | 802 | |||
BC046623.1 | - | 7 | 758 | 6315420 | ||
BC058688.1 | - | 7 | 1017 | 6413869 | ||
Y09419.1 | - | 7 | 749 |