Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam220a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026050.2 | NP_080326.2 | 3 | 1595 | ||
NM_133703.4 | NP_598464.3 | 2 | 1462 | |||
GenBank mRNA | AB374429.1 | - | 2 | 1236 | ||
AK014805.1 | - | 3 | 1528 | |||
AK075875.1 | - | 2 | 1448 | |||
AK076127.1 | - | 2 | 1445 | |||
AK156614.1 | - | 3 | 1540 | |||
AY714985.1 | - | 1 | 1011 | |||
BC005638.1 | - | 1 | 724 | |||
BC085297.1 | - | 2 | 1352 | |||
BC100440.1 | - | 2 | 1407 |