Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003008.2 | NP_002999.1 | 2 | 622 | ||
GenBank mRNA | AK312932.1 | - | 2 | 618 | ||
BC005262.1 | - | 2 | 622 | |||
BC070306.1 | - | 2 | 618 | 4356587 | ||
M81652.1 | - | 2 | 622 |