Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085486.1 | NP_001078955.1 | 4, 5 | 539 | ||
NM_001093726.1 | NP_001087195.1 | 4, 5 | 598 | |||
NM_005410.2 | NP_005401.3 | 3, 4 | 510 | |||
GenBank mRNA | AK096125.1 | - | 3, 4 | 879 | ||
AK225801.1 | - | 4, 5 | 509 | |||
AK311392.1 | - | 3, 4 | 847 | |||
AL833145.1 | - | 3, 4 | 481 | |||
AU136519.2 | - | 4, 5 | 568 | |||
BC005244.1 | - | 3, 4 | 480 | |||
BC015875.1 | - | 3, 4 | 484 | 4715833 | ||
BC040075.1 | - | 3, 4 | 460 | 5770225 | ||
BC046152.1 | - | 3, 4 | 461 | 6192976 | ||
BC058919.1 | - | 3, 4 | 491 | 6195379 | ||
Z11793.1 | - | 3, 4 | 446 |