Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETMAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243723.1 | NP_001230652.1 | 2 | 539 | ||
NM_001276325.1 | NP_001263254.1 | 2 | 539 | |||
NM_001320677.1 | NP_001307606.1 | 2 | 539 | |||
NM_006515.3 | NP_006506.3 | 2 | 539 | |||
XM_006713294.3 | XP_006713357.1 | 2 | 601 | |||
XM_017007032.1 | XP_016862521.1 | 2 | 602 | |||
GenBank mRNA | AF054989.1 | - | 2 | 465 | ||
AK222734.1 | - | 2 | 475 | |||
AK297865.1 | - | 2 | 474 | |||
AK302296.1 | - | 2 | 473 | |||
AK307537.1 | - | 2 | 539 | |||
AK311418.1 | - | 2 | 572 | |||
AY952295.1 | - | 2 | 444 | |||
BC011635.2 | - | 2 | 465 | 4080032 | ||
BG479260.1 | - | 2 | 554 | |||
U80776.1 | - | 2 | 456 |