Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015754.2 | NP_056569.2 | 5 | 470 | ||
GenBank mRNA | AK017606.1 | - | 5 | 411 | ||
AK076944.1 | - | 5 | 465 | |||
BC011107.1 | - | 5 | 408 | 3982875 | ||
BC050201.1 | - | 5 | 406 | 30013927 | ||
BC147814.1 | - | 5 | 463 | 9087531 | ||
BC147815.1 | - | 5 | 463 | 9087532 |