Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm13102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085419.1 | NP_001078888.1 | 3 | 671 | ||
GenBank mRNA | BC132592.1 | - | 3 | 672 | 40130869 | |
BC132620.1 | - | 3 | 672 | 40130897 |
Gene Symbol: Pramel4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001319.3 | NP_001001319.2 | 3 | 672 | ||
GenBank mRNA | AF490340.1 | - | 2 | 601 | ||
AK145585.1 | - | 3 | 673 | |||
AK163305.1 | - | 3 | 531 |