Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012493.2 | NP_036625.2 | 8 | 992 | ||
GenBank mRNA | BC097344.1 | - | 8 | 975 | 7459964 | |
DY317147.1 | - | 3 | 323 | |||
FQ210921.1 | - | 1 | 20 | |||
V01254.1 | - | 5 | 705 | |||
X02361.1 | - | 8 | 996 |