Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134326.2 | NP_599153.2 | 7 | 771 | ||
GenBank mRNA | BC085359.1 | - | 7 | 771 | 7303856 | |
FQ209382.1 | - | 3 | 219 | |||
FQ209651.1 | - | 11 | ||||
FQ209695.1 | - | 3 | 205 | |||
FQ209907.1 | - | 3 | 240 | |||
FQ209957.1 | - | 2 | 152 | |||
FQ210057.1 | - | 3 | 170 | |||
FQ210111.1 | - | 1 | 23 | |||
FQ210436.1 | - | 7 | 790 | |||
FQ210546.1 | - | 1 | 19 | |||
FQ210724.1 | - | 4 | 295 | |||
FQ210803.1 | - | 3 | 241 | |||
FQ210850.1 | - | 2 | 87 | |||
FQ210851.1 | - | 2 | 118 | |||
FQ210858.1 | - | 4 | 325 | |||
FQ210877.1 | - | 3 | 186 | |||
FQ211010.1 | - | 2 | 129 | |||
FQ211036.1 | - | 1 | 24 | |||
FQ211218.1 | - | 4 | 399 | |||
FQ211255.1 | - | 3 | 219 | |||
FQ211259.1 | - | 3 | 228 | |||
FQ211320.1 | - | 1 | 23 | |||
FQ218125.1 | - | 7 | 790 | |||
FQ218216.1 | - | 2 | 95 | |||
FQ218458.1 | - | 3 | 235 | |||
FQ218739.1 | - | 2 | 140 | |||
FQ218815.1 | - | 2 | 105 | |||
FQ219210.1 | - | 3 | 190 | |||
FQ219293.1 | - | 3 | 219 | |||
FQ219447.1 | - | 3 | 212 | |||
FQ219470.1 | - | 2 | 90 | |||
FQ219514.1 | - | 1 | 18 | |||
FQ219670.1 | - | 3 | 269 | |||
V01222.1 | - | 7 | 756 |