Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177305.1 | NP_001170776.1 | 7 | 796 | ||
NM_001271536.1 | NP_001258465.1 | 7 | 882 | |||
NM_012495.2 | NP_036627.1 | 7 | 898 | |||
XM_006230209.2 | XP_006230271.1 | 9 | 1144 | |||
XM_006230210.2 | XP_006230272.1 | 8 | 1054 | |||
XM_006230211.1 | XP_006230273.1 | 8 | 909 | |||
GenBank mRNA | BC064440.1 | - | 7 | 875 | 5623628 | |
FQ212782.1 | - | 7 | 849 | |||
FQ213869.1 | - | 7 | 881 | |||
FQ214123.1 | - | 7 | 849 | |||
FQ214510.1 | - | 7 | 851 | |||
FQ214648.1 | - | 3 | 270 | |||
FQ215258.1 | - | 3 | 270 | |||
FQ215311.1 | - | 3 | 270 | |||
FQ215443.1 | - | 3 | 269 | |||
FQ215968.1 | - | 3 | 270 | |||
FQ216189.1 | - | 7 | 804 | |||
FQ216413.1 | - | 3 | 271 | |||
FQ216820.1 | - | 7 | 799 | |||
FQ217110.1 | - | 3 | 270 | |||
FQ217887.1 | - | 3 | 265 | |||
FQ224097.1 | - | 3 | 270 | |||
FQ224221.1 | - | 7 | 890 | |||
FQ228954.1 | - | 7 | 828 | |||
FQ229100.1 | - | 7 | 846 | |||
M12919.1 | - | 7 | 898 | |||
M14420.1 | - | 7 | 796 |