Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177305.1 | NP_001170776.1 | 7 | 715 | ||
NM_001271536.1 | NP_001258465.1 | 7 | 801 | |||
NM_012495.2 | NP_036627.1 | 7 | 817 | |||
XM_006230209.2 | XP_006230271.1 | 9 | 1063 | |||
XM_006230210.2 | XP_006230272.1 | 8 | 973 | |||
XM_006230211.1 | XP_006230273.1 | 8 | 828 | |||
GenBank mRNA | BC064440.1 | - | 7 | 794 | 5623628 | |
CF975930.1 | - | 7 | 774 | |||
FQ213869.1 | - | 7 | 799 | |||
FQ214123.1 | - | 7 | 768 | |||
FQ214510.1 | - | 7 | 770 | |||
FQ214648.1 | - | 3 | 189 | |||
FQ215443.1 | - | 3 | 189 | |||
FQ215968.1 | - | 3 | 189 | |||
FQ216189.1 | - | 7 | 723 | |||
FQ216413.1 | - | 3 | 190 | |||
FQ216588.1 | - | 3 | 189 | |||
FQ216820.1 | - | 7 | 718 | |||
FQ217110.1 | - | 3 | 189 | |||
FQ217473.1 | - | 3 | 189 | |||
FQ217887.1 | - | 3 | 184 | |||
FQ224097.1 | - | 3 | 189 | |||
FQ228954.1 | - | 7 | 747 | |||
M12919.1 | - | 7 | 817 | |||
M14420.1 | - | 7 | 715 |