Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012506.1 | NP_036638.1 | 12 | 1625 | ||
XM_008758888.1 | XP_008757110.1 | 12 | 1539 | |||
GenBank mRNA | FQ212331.1 | - | 11 | 1462 | ||
M14513.1 | - | 12 | 1625 | |||
M28648.1 | - | 11 | 1479 | |||
X05883.1 | - | 12 | 1622 |