Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012507.3 | NP_036639.2 | 2, 3 | 822 | ||
XM_006246580.2 | XP_006246642.1 | 2, 3 | 845 | |||
XM_006246581.2 | XP_006246643.1 | 2, 3 | 845 | |||
GenBank mRNA | BC087034.1 | - | 2, 3 | 822 | 7124310 | |
J04629.1 | - | 2, 3 | 711 | |||
U45946.1 | - | 2, 3 | 260 |