Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012507.3 | NP_036639.2 | 6, 7 | 1274 | ||
XM_006246580.2 | XP_006246642.1 | 8, 9 | 1345 | |||
XM_006246581.2 | XP_006246643.1 | 7, 8 | 1324 | |||
GenBank mRNA | BC087034.1 | - | 6, 7 | 1274 | 7124310 | |
J04629.1 | - | 6, 7 | 1163 | |||
U45946.1 | - | 6, 7 | 712 |