Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012510.2 | NP_036642.2 | 2 | 305 | ||
XM_008771385.1 | XP_008769607.1 | 2 | 130 | |||
GenBank mRNA | M35535.1 | - | 2 | 305 | ||
M55655.1 | - | 2 | 141 |