Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270630.1 | NP_001257559.1 | 2 | 915 | ||
NM_001270631.1 | NP_001257560.1 | 2 | 1066 | |||
NM_001270632.1 | NP_001257561.1 | 2 | 983 | |||
NM_001270633.1 | NP_001257562.1 | 2 | 907 | |||
NM_001270634.1 | NP_001257563.1 | 3 | 794 | |||
NM_001270635.1 | NP_001257564.1 | 2 | 793 | |||
NM_001270636.1 | NP_001257565.1 | 2 | 779 | |||
NM_001270637.1 | NP_001257566.1 | 2 | 771 | |||
NM_001270638.1 | NP_001257567.1 | 2 | 648 | |||
NM_012513.4 | NP_036645.2 | 2 | 1206 | |||
XM_006234684.2 | XP_006234746.1 | 2 | 914 | |||
XM_008762078.1 | XP_008760300.1 | 2 | 2840 | |||
GenBank mRNA | AY559248.1 | - | 2 | 709 | ||
AY559249.1 | - | 2 | 723 | |||
AY559250.1 | - | 3 | 795 | |||
BC087634.1 | - | 2 | 901 | 7319966 | ||
EF125675.1 | - | 2 | 1206 | |||
EF125676.1 | - | 2 | 772 | |||
EF125677.1 | - | 2 | 984 | |||
EF125678.1 | - | 2 | 1067 | |||
EF125679.1 | - | 2 | 907 | |||
EF125680.1 | - | 2 | 915 | |||
EF125686.1 | - | 2 | 793 | |||
EF125687.1 | - | 2 | 648 | |||
EF125688.1 | - | 2 | 780 | |||
EF125689.1 | - | 2 | 844 | |||
EF125690.1 | - | 1 | 1038 | |||
KC855561.1 | - | 1 | 76 | |||
M61175.1 | - | 2 | 618 | |||
X67108.1 | - | 1 | 567 |