Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243879.1 | NP_001230808.1 | 5 | 849 | ||
NM_004593.2 | NP_004584.1 | 6 | 983 | |||
XM_005247703.1 | XP_005247760.1 | 7 | 1151 | |||
XM_011513072.2 | XP_011511374.1 | 7 | 3953 | |||
XM_017007033.1 | XP_016862522.1 | 6 | 2129 | |||
GenBank mRNA | AB209208.1 | - | 7 | 3940 | ||
AK091863.1 | - | 2 | 1870 | |||
AK094495.1 | - | 3 | 905 | |||
AK098191.1 | - | 6 | 770 | |||
AK125610.1 | - | 3 | 1653 | |||
AK301118.1 | - | 5 | 702 | |||
AK315632.1 | - | 6 | 967 | |||
BC000160.2 | - | 6 | 850 | 2984771 | ||
BC000451.2 | - | 6 | 849 | 2821443 | ||
BC005898.1 | - | 6 | 834 | 4046349 | ||
CR749214.1 | - | 5 | 3010 | |||
U61267.1 | - | 6 | 828 | |||
U68063.1 | - | 6 | 836 |