Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031969.2 | NP_114175.1 | 3 | 171 | ||
GenBank mRNA | AF178845.1 | - | 106 | |||
FQ211760.1 | - | 3 | 238 | |||
FQ212721.1 | - | 576 | ||||
FQ213227.1 | - | 3 | 284 | |||
FQ225380.1 | - | 3 | 197 | |||
FQ226971.1 | - | 3 | 243 | |||
FQ227186.1 | - | 3 | 243 | |||
X13933.1 | - | 3 | 171 |