Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012520.2 | NP_036652.1 | 8 | 1058 | ||
GenBank mRNA | BC081853.1 | - | 8 | 955 | 7109889 | |
M11670.1 | - | 8 | 1009 |