Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Comt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012531.2 | NP_036663.1 | 2 | 424 | ||
XM_006248603.2 | XP_006248665.1 | 2 | 425 | |||
GenBank mRNA | BC081850.1 | - | 2 | 282 | 7191956 | |
CK478774.1 | - | 2 | 544 | |||
FQ209401.1 | - | 1 | 137 | |||
FQ209488.1 | - | 1 | 143 | |||
FQ209638.1 | - | 1 | 137 | |||
FQ209670.1 | - | 1 | 137 | |||
FQ209706.1 | - | 1 | 148 | |||
FQ210267.1 | - | 1 | 137 | |||
FQ210416.1 | - | 1 | 173 | |||
FQ210430.1 | - | 1 | 140 | |||
FQ218190.1 | - | 1 | 139 | |||
FQ218375.1 | - | 1 | 147 | |||
FQ218564.1 | - | 1 | 139 | |||
FQ218595.1 | - | 1 | 139 | |||
FQ219155.1 | - | 1 | 144 | |||
FQ219257.1 | - | 1 | 144 | |||
FQ219433.1 | - | 1 | 139 | |||
FQ223536.1 | - | 1 | 139 | |||
FQ228829.1 | - | 1 | 137 | |||
FQ228870.1 | - | 1 | 145 | |||
FQ229523.1 | - | 1 | 119 | |||
M60753.1 | - | 228 |