Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crygd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033095.1 | NP_149086.1 | 3 | 393 | ||
XM_008767074.1 | XP_008765296.1 | 2 | 1264 | |||
GenBank mRNA | X57169.1 | - | 3 | 378 |