Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl3d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033233.1 | NP_150236.1 | 4 | 452 | ||
XM_008771595.1 | XP_008769817.1 | 4 | 434 | |||
GenBank mRNA | BC097302.1 | - | 4 | 478 | 7444366 | |
U32679.1 | - | 4 | 452 |
Gene Symbol: LOC100911206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257188.2 | XP_006257250.2 | 1 | 32 |
Gene Symbol: LOC103689989 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771673.1 | XP_008769895.1 | 434 | |||
XM_008771674.1 | XP_008769896.1 | 472 |
Gene Symbol: LOC103690150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771672.1 | XP_008769894.1 | 469 | |||
GenBank mRNA | FQ228618.1 | - | 492 |
Gene Symbol: Prl3d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253914.2 | 4 | 492 | |||
GenBank mRNA | FQ220053.1 | - | 496 | |||
FQ220072.1 | - | 492 | ||||
FQ220274.1 | - | 494 | ||||
FQ220568.1 | - | 494 | ||||
FQ220587.1 | - | 492 | ||||
FQ220864.1 | - | 218 | ||||
FQ221231.1 | - | 494 | ||||
FQ222593.1 | - | 492 | ||||
FQ223300.1 | - | 495 | ||||
FQ228787.1 | - | 492 | ||||
FQ229328.1 | - | 492 | ||||
FQ229537.1 | - | 491 | ||||
FQ229656.1 | - | 486 | ||||
FQ230119.1 | - | 492 | ||||
M55269.1 | - | 485 |
Gene Symbol: Prl3d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771671.1 | XP_008769893.1 | 4 | 495 | ||
GenBank mRNA | BC161851.1 | - | 4 | 469 | 9088452 | |
D21103.1 | - | 4 | 431 | |||
DQ329282.1 | - | 4 | 488 | |||
DQ329283.1 | - | 4 | 488 | |||
FQ145043.1 | - | 4 | 494 | |||
FQ221365.1 | - | 4 | 492 | |||
FQ223255.1 | - | 4 | 495 | |||
FQ223518.1 | - | 4 | 492 | |||
FQ229928.1 | - | 4 | 494 | |||
FQ230035.1 | - | 4 | 492 |