Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012536.1 | NP_036668.1 | 4, 5 | 313 | ||
GenBank mRNA | FQ232114.1 | - | 4, 5 | 335 | ||
FQ233595.1 | - | 4, 5 | 335 | |||
FQ234337.1 | - | 4, 5 | 335 | |||
FQ234339.1 | - | 4, 5 | 335 |