Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ace | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012544.1 | NP_036676.1 | 3884 | |||
GenBank mRNA | AF201331.1 | - | 3884 | |||
AF201332.1 | - | 3884 | ||||
AF532783.1 | - | 3828 | ||||
AF532784.1 | - | 3828 | ||||
AF539425.1 | - | 2261 | ||||
AF539701.1 | - | 319 | ||||
BC085760.1 | - | 3884 | 7097136 | |||
U03708.1 | - | 3884 | ||||
U03734.1 | - | 3884 |
Gene Symbol: LOC102556346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247643.2 | XP_006247705.1 | 3 | 1237 |