Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nqo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017000.3 | NP_058696.2 | 2 | 212 | ||
GenBank mRNA | BC083542.1 | - | 2 | 212 | 7190779 | |
J02608.1 | - | 2 | 197 | |||
J02640.1 | - | 1 | 171 | |||
J02679.1 | - | 2 | 197 |