Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000337.5 | NP_000328.2 | 6 | 913 | ||
NM_001128209.1 | NP_001121681.1 | 5 | 867 | |||
NM_172244.2 | NP_758447.1 | 6 | 913 | |||
XM_005265966.4 | XP_005266023.1 | 6 | 741 | |||
XM_005265967.2 | XP_005266024.1 | 6 | 505 | |||
XM_011534621.2 | XP_011532923.1 | 5 | 570 | |||
XM_017009723.1 | XP_016865212.1 | 7 | 679 | |||
XM_017009724.1 | XP_016865213.1 | 7 | 956 | |||
GenBank mRNA | AK292794.1 | - | 6 | 505 | ||
BC008431.1 | - | 8 | 812 | |||
BC020740.1 | - | 6 | 506 | 4702959 | ||
BX537948.1 | - | 5 | 878 | |||
DA561069.1 | - | 5 | 460 | |||
U58331.1 | - | 5 | 432 | |||
X95191.1 | - | 4 | 394 |