Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017002.2 | NP_058698.2 | 5 | 735 | ||
XM_006242602.2 | XP_006242664.1 | 5 | 841 | |||
XM_006242603.2 | XP_006242665.1 | 5 | 1338 | |||
GenBank mRNA | BC089810.1 | - | 5 | 735 | 7375416 | |
D13566.1 | - | 5 | 653 | |||
D83509.1 | - | 5 | 653 |