Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017005.2 | NP_058701.2 | 10 | 1441 | ||
GenBank mRNA | BC087598.1 | - | 10 | 1441 | 7311498 | |
FQ218764.1 | - | 6 | 752 | |||
FQ230216.1 | - | 6 | 786 | |||
FQ232207.1 | - | 6 | 786 | |||
J04473.1 | - | 10 | 1452 |