Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144757.1 | NP_001138229.1 | 5, 6 | 664 | ||
NM_003020.3 | NP_003011.1 | 5, 6 | 661 | |||
GenBank mRNA | AK314965.1 | - | 5, 6 | 657 | ||
BC005349.1 | - | 5, 6 | 603 | 3949961 | ||
BC093053.1 | - | 5, 6 | 620 | 30720075 | ||
CR541824.1 | - | 4, 5 | 544 | |||
Y00757.1 | - | 5, 6 | 572 |