Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003021.3 | NP_003012.1 | 4, 5 | 444 | ||
XM_011528178.2 | XP_011526480.1 | 4, 5 | 498 | |||
GenBank mRNA | AF368279.1 | - | 3, 4 | 318 | ||
AF408399.1 | - | 3, 4 | 297 | |||
AJ133129.1 | - | 4, 5 | 339 | |||
AJ223828.1 | - | 3, 4 | 338 | |||
AK293535.1 | - | 4, 5 | 415 | |||
AL050156.1 | - | 4, 5 | 406 | |||
BC000390.2 | - | 4, 5 | 347 | 2821268 | ||
BC002989.1 | - | 4, 5 | 347 | 2821268 | ||
BC005165.2 | - | 4, 5 | 341 | 3532317 | ||
BC008885.2 | - | 4, 5 | 341 | 4302652 | ||
BG423904.1 | - | 445 | ||||
CR533517.1 | - | 3, 4 | 297 | |||
CR542282.1 | - | 3, 4 | 297 |