Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012574.1 | NP_036706.1 | 6 | 1702 | ||
GenBank mRNA | M91562.1 | - | 6 | 1409 | ||
U11419.1 | - | 6 | 1702 |