Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003021.3 | NP_003012.1 | 5 | 483 | ||
XM_011528178.2 | XP_011526480.1 | 5 | 537 | |||
GenBank mRNA | AF368279.1 | - | 4 | 357 | ||
AF408399.1 | - | 4 | 336 | |||
AJ133129.1 | - | 5 | 378 | |||
AJ223828.1 | - | 4 | 377 | |||
AK293535.1 | - | 5 | 454 | |||
AL050156.1 | - | 5 | 445 | |||
BC000390.2 | - | 5 | 386 | 2821268 | ||
BC002989.1 | - | 5 | 386 | 2821268 | ||
BC005165.2 | - | 5 | 380 | 3532317 | ||
BC008885.2 | - | 5 | 380 | 4302652 | ||
BG423904.1 | - | 484 | ||||
CR533517.1 | - | 4 | 336 | |||
CR542282.1 | - | 4 | 336 |