Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105711.1 | NP_001099181.1 | 4 | 1561 | ||
GenBank mRNA | M92075.1 | - | 5 | 1554 |
Set Membership: |
G-protein coupled receptor |