Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017013.4 | NP_058709.2 | 3 | 162 | ||
GenBank mRNA | BC078706.1 | - | 3 | 154 | 7128611 | |
FJ179397.1 | - | 2 | 99 |