Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017014.1 | NP_058710.1 | 5 | 373 | ||
GenBank mRNA | BC063172.1 | - | 5 | 413 | 6920688 | |
FJ179401.1 | - | 5 | 337 | |||
FQ211067.1 | - | 5 | 372 | |||
FQ212742.1 | - | 5 | 366 | |||
FQ213204.1 | - | 5 | 366 | |||
FQ213696.1 | - | 5 | 560 | |||
FQ214055.1 | - | 5 | 472 | |||
FQ217092.1 | - | 5 | 439 | |||
FQ218381.1 | - | 5 | 373 | |||
FQ218789.1 | - | 5 | 372 | |||
FQ218795.1 | - | 5 | 409 | |||
FQ229836.1 | - | 5 | 420 | |||
FQ229941.1 | - | 5 | 400 | |||
J02810.1 | - | 5 | 373 | |||
M11719.1 | - | 5 | 374 | |||
M28241.1 | - | 5 | 433 | |||
X04229.1 | - | 5 | 373 |