Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1f0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012578.2 | NP_036710.1 | 1 | 380 | ||
GenBank mRNA | BC061842.1 | - | 1 | 380 | 5621199 | |
X70685.1 | - | 1 | 150 | |||
X72624.1 | - | 1 | 258 |