Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033234.1 | NP_150237.1 | 3 | 375 | ||
GenBank mRNA | BC058448.1 | - | 3 | 384 | 6921704 | |
M17084.1 | - | 3 | 332 | |||
X16417.1 | - | 3 | 375 |
Gene Symbol: Hbb-b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198776.1 | NP_942071.1 | 3 | 384 | ||
XM_006229913.2 | XP_006229975.2 | 4 | 456 | |||
GenBank mRNA | BC058502.1 | - | 3 | 384 | 6887460 | |
FQ218177.1 | - | 3 | 378 |
Gene Symbol: LOC100134871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113223.1 | NP_001106694.1 | 3 | 379 | ||
GenBank mRNA | FQ209951.1 | - | 3 | 382 | ||
FQ210023.1 | - | 3 | 382 | |||
FQ210060.1 | - | 3 | 380 | |||
FQ210106.1 | - | 3 | 380 | |||
FQ210483.1 | - | 3 | 380 | |||
FQ210600.1 | - | 3 | 382 | |||
FQ210669.1 | - | 3 | 376 | |||
FQ210740.1 | - | 3 | 380 | |||
FQ211039.1 | - | 3 | 374 | |||
FQ211266.1 | - | 3 | 380 | |||
FQ211272.1 | - | 3 | 380 | |||
FQ211275.1 | - | 3 | 380 | |||
FQ211294.1 | - | 3 | 382 | |||
FQ211362.1 | - | 3 | 380 | |||
FQ211415.1 | - | 3 | 380 | |||
FQ212253.1 | - | 3 | 380 | |||
FQ212406.1 | - | 3 | 382 | |||
FQ212421.1 | - | 3 | 380 | |||
FQ213080.1 | - | 3 | 409 | |||
FQ214057.1 | - | 3 | 380 | |||
FQ214129.1 | - | 3 | 380 | |||
FQ217131.1 | - | 3 | 378 | |||
FQ217673.1 | - | 3 | 380 | |||
FQ218002.1 | - | 3 | 381 | |||
FQ218191.1 | - | 3 | 380 | |||
FQ218246.1 | - | 3 | 352 | |||
FQ218266.1 | - | 3 | 418 | |||
FQ218689.1 | - | 3 | 380 | |||
FQ218821.1 | - | 3 | 380 | |||
FQ219109.1 | - | 3 | 380 | |||
FQ219133.1 | - | 3 | 411 | |||
FQ221588.1 | - | 3 | 378 | |||
FQ222844.1 | - | 3 | 380 | |||
FQ224149.1 | - | 3 | 380 | |||
FQ224301.1 | - | 3 | 380 | |||
FQ224598.1 | - | 3 | 380 | |||
FQ224632.1 | - | 3 | 380 | |||
FQ224738.1 | - | 3 | 380 | |||
FQ225377.1 | - | 3 | 380 | |||
FQ226460.1 | - | 3 | 380 | |||
FQ226535.1 | - | 3 | 380 | |||
FQ226955.1 | - | 3 | 380 | |||
FQ228555.1 | - | 3 | 380 | |||
FQ232298.1 | - | 524 | ||||
FQ232442.1 | - | 3 | 411 | |||
FQ233001.1 | - | 3 | 380 | |||
FQ234495.1 | - | 3 | 381 | |||
X05080.1 | - | 3 | 379 | |||
X16418.1 | - | 3 | 345 |
Gene Symbol: LOC103694855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773799.1 | XP_008772021.1 | 3 | 420 | ||
XM_008773800.1 | XP_008772022.1 | 4 | 452 |
Gene Symbol: LOC103694857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773801.1 | XP_008772023.1 | 3 | 416 |
Gene Symbol: LOC689064 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111269.1 | NP_001104739.1 | 362 | |||
GenBank mRNA | DY319996.1 | - | 362 | |||
FQ209804.1 | - | 382 | ||||
FQ209902.1 | - | 382 | ||||
FQ210004.1 | - | 379 | ||||
FQ210039.1 | - | 382 | ||||
FQ210298.1 | - | 380 | ||||
FQ210379.1 | - | 382 | ||||
FQ210599.1 | - | 380 | ||||
FQ210742.1 | - | 380 | ||||
FQ210889.1 | - | 382 | ||||
FQ211252.1 | - | 380 | ||||
FQ211410.1 | - | 380 | ||||
FQ211471.1 | - | 379 | ||||
FQ211709.1 | - | 455 | ||||
FQ215055.1 | - | 379 | ||||
FQ217121.1 | - | 411 | ||||
FQ217477.1 | - | 380 | ||||
FQ217746.1 | - | 380 | ||||
FQ218196.1 | - | 380 | ||||
FQ218287.1 | - | 380 | ||||
FQ218323.1 | - | 380 | ||||
FQ221211.1 | - | 380 | ||||
FQ225430.1 | - | 380 | ||||
FQ225765.1 | - | 380 | ||||
FQ226110.1 | - | 380 | ||||
FQ228130.1 | - | 910 | ||||
FQ230470.1 | - | 382 | ||||
FQ230522.1 | - | 387 | ||||
FQ230899.1 | - | 384 | ||||
FQ231257.1 | - | 196 |