Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hprt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012583.2 | NP_036715.1 | 3 | 303 | ||
GenBank mRNA | BC098629.1 | - | 3 | 303 | 7386548 | |
M63983.1 | - | 3 | 335 | |||
M86443.1 | - | 63 | ||||
S79292.1 | - | 3 | 335 | |||
X62085.1 | - | 263 |
Gene Symbol: LOC103689983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773659.1 | XP_008771881.1 | 3 | 379 |