Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122681.1 | NP_001116153.1 | 8 | 1114 | ||
NM_001145855.1 | NP_001139327.1 | 8 | 1257 | |||
NM_001145856.1 | NP_001139328.1 | 8 | 1204 | |||
NM_003023.4 | NP_003014.3 | 8 | 1255 | |||
GenBank mRNA | AB000462.1 | - | 8 | 1255 | ||
AB000463.1 | - | 9 | 1434 | |||
AF000936.1 | - | 7 | 994 | |||
AK124073.1 | - | 3 | 1100 | |||
AK126537.1 | - | 3 | 1471 | |||
AK225497.1 | - | 8 | 1116 | |||
AK299996.1 | - | 8 | 1173 | |||
AK312286.1 | - | 8 | 1052 | |||
BC022996.1 | - | 8 | 1110 | 5103986 | ||
U56386.1 | - | 8 | 1202 |