Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122681.1 | NP_001116153.1 | 11 | 1575 | ||
NM_001145855.1 | NP_001139327.1 | 11 | 1718 | |||
NM_001145856.1 | NP_001139328.1 | 11 | 1665 | |||
NM_003023.4 | NP_003014.3 | 11 | 1716 | |||
GenBank mRNA | AB000462.1 | - | 11 | 1716 | ||
AB000463.1 | - | 12 | 1895 | |||
AF000936.1 | - | 10 | 1455 | |||
AK124073.1 | - | 6 | 1561 | |||
AK126537.1 | - | 5 | 2284 | |||
AK225497.1 | - | 11 | 1577 | |||
AK299996.1 | - | 11 | 1634 | |||
AK312286.1 | - | 11 | 1513 | |||
BC022996.1 | - | 11 | 1571 | 5103986 | ||
U56386.1 | - | 11 | 1663 |