Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082477.2 | NP_001075946.2 | 3 | 324 | ||
NM_001082478.1 | NP_001075947.1 | 3 | 621 | |||
NM_001082479.1 | NP_001075948.1 | 3 | 621 | |||
NM_178866.4 | NP_849197.3 | 3 | 324 | |||
XM_006241191.2 | XP_006241253.1 | 4 | 465 | |||
XM_006241192.1 | XP_006241254.1 | 4 | 379 | |||
XM_006241193.2 | XP_006241255.1 | 4 | 464 | |||
XM_006241194.2 | XP_006241256.1 | 3 | 503 | |||
GenBank mRNA | BC086374.1 | - | 3 | 324 | 7300903 | |
D00698.1 | - | 3 | 305 | |||
FQ209824.1 | - | 4 | 453 | |||
FQ210560.1 | - | 3 | 337 | |||
FQ210688.1 | - | 3 | 337 | |||
FQ220385.1 | - | 3 | 645 | |||
FQ225388.1 | - | 3 | 337 | |||
FQ225437.1 | - | 3 | 337 | |||
FQ231951.1 | - | 3 | 337 | |||
FQ234943.1 | - | 3 | 337 | |||
M17335.1 | - | 3 | 521 | |||
X06043.1 | - | 2 | 206 | |||
X06107.1 | - | 3 | 599 | |||
X06108.1 | - | 4 | 413 |