Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031513.1 | NP_113701.1 | 3, 4 | 343 | ||
XM_006246226.2 | XP_006246288.1 | 3, 4 | 319 | |||
GenBank mRNA | U81492.1 | - | 3, 4 | 343 | ||
U81493.1 | - | 3, 4 | 334 |