Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jun | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021835.3 | NP_068607.1 | 1 | 1675 | ||
GenBank mRNA | BC078738.1 | - | 1 | 912 | 7124370 | |
X17163.1 | - | 1 | 1169 |