Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017023.1 | NP_058719.1 | 2 | 332 | ||
XM_008766044.1 | XP_008764266.1 | 3 | 463 | |||
XM_008766045.1 | XP_008764267.1 | 3 | 1153 | |||
XM_008766046.1 | XP_008764268.1 | 2 | 341 | |||
XM_008766047.1 | XP_008764269.1 | 2 | 454 | |||
XM_008766048.1 | XP_008764270.1 | 2 | 1031 | |||
GenBank mRNA | AF081365.1 | - | 2 | 332 | ||
AF081366.1 | - | 2 | 281 | |||
AF081367.1 | - | 3 | 299 | |||
AF081368.1 | - | 3 | 454 | |||
L29403.1 | - | 2 | 281 | |||
X72341.1 | - | 2 | 332 |