Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012600.2 | NP_036732.2 | 14 | 1661 | ||
XM_008766439.1 | XP_008764661.1 | 14 | 1958 | |||
GenBank mRNA | CO395247.1 | - | 2 | 172 | ||
FQ228526.1 | - | 14 | 1692 |