Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012605.2 | NP_036737.1 | 2 | 86 | ||
GenBank mRNA | CB770854.1 | - | 2 | 96 | ||
FQ215085.1 | - | 2 | 119 | |||
FQ216891.1 | - | 2 | 102 | |||
FQ216934.1 | - | 2 | 102 | |||
FQ217090.1 | - | 2 | 104 | |||
FQ217098.1 | - | 2 | 102 | |||
FQ217155.1 | - | 2 | 102 | |||
FQ217219.1 | - | 2 | 102 | |||
FQ217259.1 | - | 2 | 102 | |||
FQ217349.1 | - | 1 | 26 | |||
FQ217365.1 | - | 2 | 101 | |||
FQ217368.1 | - | 2 | 118 | |||
FQ217533.1 | - | 2 | 130 | |||
FQ217703.1 | - | 2 | 102 | |||
FQ217740.1 | - | 2 | 118 | |||
FQ217881.1 | - | 2 | 99 | |||
FQ217944.1 | - | 2 | 102 | |||
FQ218017.1 | - | 2 | 102 | |||
FQ223762.1 | - | 2 | 83 | |||
FQ223868.1 | - | 2 | 102 | |||
FQ223911.1 | - | 2 | 118 | |||
FQ223946.1 | - | 2 | 102 | |||
FQ223966.1 | - | 2 | 102 | |||
FQ223985.1 | - | 2 | 102 | |||
FQ224231.1 | - | 2 | 102 | |||
FQ224391.1 | - | 2 | 100 | |||
FQ224404.1 | - | 2 | 100 | |||
FQ224511.1 | - | 2 | 102 | |||
FQ224525.1 | - | 2 | 121 | |||
FQ224720.1 | - | 2 | 102 | |||
FQ224781.1 | - | 2 | 118 |