Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012611.3 | NP_036743.3 | 2441 | |||
GenBank mRNA | AF006619.1 | - | 9 | 978 | ||
AF006620.1 | - | 9 | 978 | |||
AY211532.1 | - | 2252 | ||||
D12520.1 | - | 2441 | ||||
D14051.1 | - | 2271 | ||||
D44591.1 | - | 2272 | ||||
D83661.1 | - | 2251 | ||||
L12562.1 | - | 2353 | ||||
U03699.1 | - | 2254 | ||||
U26686.1 | - | 2251 | ||||
X76881.1 | - | 2436 |
Gene Symbol: LOC497963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246949.2 | XP_006247011.1 | 2306 |