Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017031.2 | NP_058727.2 | 7, 8 | 1301 | ||
GenBank mRNA | AF202732.1 | - | 1159 | |||
AF202733.1 | - | 7, 8 | 865 | |||
J04563.1 | - | 4, 5 | 313 | |||
L27058.1 | - | 2, 3 | 860 | |||
U95748.1 | - | 7, 8 | 1301 |